

Lucigenin ≥97.0% (by HPLC) for chemiluminescence research
Supplier: TCI
Certificates
About this item
CAS Number:2315-97-1
:22103-92-0
Formula:C₂₈H₂₂N₄O₆
MDL Number:MFCD00011925
:510.51 g/mol
EINECS:219-023-4
UN:1479
ADR:5.1,III
:GHS03
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 97.0 area%
- Molar extinction coefficient:min. 150000 (H2O, 260.0 to 261.0 nm)
- Molar extinction coefficient2:min. 33000 (H2O, 368.0 to 369.0 nm)
Specification Test Results
Frequently Bought Together