Ethyl-2-butylacetoacetate ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 2-Acetylhexanoic Acid Ethyl Ester, 2-Butylacetoacetic Acid Ethyl Ester, Ethyl 2-acetylhexanoate, Ethyl 2-Acetylhexanoate
CAS Number:1540-29-0
:
Formula:C₁₀H₁₈O₃
MDL Number:MFCD00026888
Boiling Pt:224°C
Molecular Weight:186.25 g/mol
EINECS:216-271-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together