Trifluoroacetic acid ∼0.5 mol/L in water for ion pair chromatography, for LC-MS
Catalog # TCIAA5711-10ML
Supplier: TCI
CAS Number:
Specifications
- Pk:10 mL
- Seq:2275
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Concentration (Neutralization titration):0.50 to 0.55 mol/L
- Absorbance (240nm):max. 1.00
- Absorbance (254nm):max. 0.15
- Absorbance (280nm):max. 0.05
Specification Test Results
About this item
:
:114.02 g/mol
Merck Index:14,09681
EINECS:200-929-3
UN:3265
ADR:8,II
:GHS05
CAS Number:76-05-1
Formula:C₂HF₃O₂
MDL Number:MFCD00004169