6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 6-(4-Aminophenyl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, Levosimendan dazinone intermediate
CAS Number:36725-28-7
:
Formula:C₁₁H₁₃N₃O
MDL Number:MFCD01692712
:203.25 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together