Argatroban monohydrate ≥99.0% (by HPLC)
TCIAA2705-25MG
: TCI
:
- Pk:25 mg
- Seq:2315
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 99.0 area%
- Elemental analysis (Nitrogen):15.50 to 16.50 %
- Specific rotation [a]20/D:+160.0 to +170.0 deg (C=0.8, MeOH) (calcd.on anh.substance)
- Water:2.5 to 4.5 %
:
:526.65 g/mol
:14,00779
:180°C
:Refrigerator
:141396-28-3
:C₂₃H₃₆N₆O₅S·H₂O
:MFCD00895735