4-Amino-N-(tert-butoxycarbonyl)-D-phenylalanine ≥97.0% (par HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:164332-89-2
:
Formula:C₁₄H₂₀N₂O₄
MDL Number:MFCD00236818
:280.32 g/mol
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (HPLC):min. 97.0 area%
- Purity (Nonaqueous Titration):min. 95.0 %
- Specific rotation [a]20/D:-23.0 to -27.0 deg (C=1, methanol)
Specification Test Results
Frequently Bought Together