Order Entry
Belgium
ContactUsLinkComponent
5-Azacytidine ≥98.0% (by titrimetric analysis)
5-Azacytidine ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
5-Azacytidine ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together