Potassium-L-aspartate hydrate ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1115-63-5
:
Formula:C₄H₆KNO₄·xH₂O
MDL Number:MFCD00021083
:171.19 g/mol (anhy)
Merck Index:14,00840
EINECS:214-226-4
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:+19.0 to +22.0 deg (C=8, 6mol/L HCl) (calcd.on anh.substance)
- Water:max. 6.0 %
Specification Test Results
Frequently Bought Together