N-Acetyl-DL-norleucine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
Synonyms: Ac-DL-Nle-OH
CAS Number:7682-16-8
:
Formula:C₈H₁₅NO₃
MDL Number:MFCD00037272
:173.21 g/mol
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:106.0 to 109.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together