
meta-Phosphoric acid stabilised, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:37267-86-0
:
Formula:(HPO₃)n
MDL Number:MFCD00011351
:79.98 g/mol
Boiling Pt:260 °C (1013 hPa)
Melting Pt:21 °C
Density:2.4 g/cm³ (20 °C)
EINECS:253-433-4
UN:3453
ADR:8,III
:M-Clarity™ quality level = MQ300
:Stabilised with sodium metaphosphate
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble,Hydrolysis
- Melting Point:200 - 250 °C
- Bulk Density:1000 kg/m3
- Boiling Point:600 °C (1013 hP)
- Vapor Pressure:- 0.001 hPa (20 °C)
- Density:2.0 g/cm3 (20 °C)
- pH:2 (33 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Frequently Bought Together