Ammonium heptamolybdate tetrahydrate 81-83% (MoO₃ basis) ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12054-85-2
:12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059
:1235.86 g/mol
Merck Index:14,533
Melting Pt:∼ 190 °C (dec.)
Density:2.498
EINECS:234-722-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together