Magnesium hydrogen orthophosphate trihydrate
Catalog # LOHM303574.9025
Supplier: LOHMANN
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:174.33 g/mol
Density:2.131 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-823-5
CAS Number:7782-75-4
Formula:HMgO₄P·3H₂O
MDL Number:MFCD00150390