Order Entry
Belgium
ContactUsLinkComponent
Magnesium hydrogen orthophosphate trihydrate
Magnesium hydrogen orthophosphate trihydrate
Catalog # LOHM303573.9025
Supplier:  LOHMANN
CAS Number:  
Magnesium hydrogen orthophosphate trihydrate
Catalog # LOHM303573.9025
Supplier:  LOHMANN
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :174.33 g/mol
Density:2.131 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-823-5
CAS Number:7782-75-4
Formula:HMgO₄P·3H₂O
MDL Number:MFCD00150390