Order Entry
Belgium
ContactUsLinkComponent
D-(-)-N-Methylglucamine
D-(-)-N-Methylglucamine
Catalog # MOLE16262469
Supplier:  Molekula
CAS Number:  
D-(-)-N-Methylglucamine
Catalog # MOLE16262469
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :195.22 g/mol
Merck Index:12,06154
Melting Pt:128…131 °C
Storage Temperature:Ambient
EINECS:228-506-9
CAS Number:6284-40-8
Formula:H₃CNHCH₂(CH(OH))₄CH₂OH
MDL Number:MFCD00004707