Order Entry
Belgium
ContactUsLinkComponent
D-(-)-N-Methylglucamine ≥99.0% for resolution of racemates, for synthesis, Sigma-Aldrich®
D-(-)-N-Methylglucamine ≥99.0% for resolution of racemates, for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
D-(-)-N-Methylglucamine ≥99.0% for resolution of racemates, for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    1000 g/l (25 °C)
  • Melting Point:
    128 - 131 °C
  • Molar Mass:
    195.21 g/mol
  • Bulk Density:
    300 kg/m3
  • Boiling Point:
    210 °C (1013 hPa) decomposes
  • pH:
    11 (10 g/l, H2O, 20 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together