

Dodecamolybdophosphoric acid hydrate, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:51429-74-4
:
Formula:H₃[P(Mo₃O₁₀)₄]·nH₂O
MDL Number:MFCD00149913
:1843.27 g/mol
Merck Index:12,07498
Melting Pt:78…90 °C
Density:1.62 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:234-713-5
UN:3260
ADR:8,III
:GHS03
:GHS05
Frequently Bought Together