Order Entry
Belgium
ContactUsLinkComponent
Dodecamolybdophosphoric acid hydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Dodecamolybdophosphoric acid hydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Dodecamolybdophosphoric acid hydrate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    78 - 90 °C (anhydrous substance)
  • Molar Mass:
    1825.28 g/mol (anhydrous substance)
  • Bulk Density:
    980 kg/m3
  • pH:
    (H2O) strongly acid

Specification Test Results

Related Information

Frequently Bought Together