p-Toluenesulphonic acid monohydrate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
Synonyms: 4-Methylbenzenesulphonic acid hydrate, Toluene-4-sulphonic acid monohydrate
CAS Number:6192-52-5
:
Formula:C₇H₈O₃S·1H₂O
MDL Number:MFCD00064387
:190.22 g/mol
Merck Index:14,09533
Melting Pt:102…104 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:180 °C
Storage Temperature:Ambient
EINECS:203-180-0
UN:2585
ADR:8,III
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):98.0 to 102.0 % (after drying)
- Melting point:104.0 to 107.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together