L(+)-Lysine monohydrochloride
Supplier: Serva
Certificates
About this item
CAS Number:657-27-2
:
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564
Melting Pt:263…264 °C
:182.65 g/mol
Storage Temperature:Ambient
EINECS:211-519-9
Frequently Bought Together