Order Entry
Belgium
ContactUsLinkComponent
L(+)-Lysine monohydrochloride
L(+)-Lysine monohydrochloride
Catalog # A1706.1000
Supplier:  PanReac AppliChem
CAS Number:  
L(+)-Lysine monohydrochloride
Catalog # A1706.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Melting Pt:263…264 °C
 :182.65 g/mol
Storage Temperature:Ambient
EINECS:211-519-9
CAS Number:657-27-2
Formula:H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MDL Number:MFCD00064564