Order Entry
Belgium
ContactUsLinkComponent
Magnesium sulphate 1 mol/l in aqueous solution
Magnesium sulphate 1 mol/l in aqueous solution
Catalog # ROCKMB-056
CAS Number:  
undefined
Magnesium sulphate 1 mol/l in aqueous solution
Catalog # ROCKMB-056
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :120.37 g/mol
Storage Temperature:Ambient
EINECS:231-298-2
CAS Number:7487-88-9
Formula:MgSO₄
MDL Number:MFCD00011110