Order Entry
Belgium
ContactUsLinkComponent
 
 
Barium chloride 0.1 mol/l (0.2 N) in aqueous solution
  FFCHFF20480500
 :  
 
Barium chloride 0.1 mol/l (0.2 N) in aqueous solution
  FFCHFF20480500
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

 :
 :208.24 g/mol
 :1.034 g/cm³ (20 °C)
 :Ambient
 :233-788-1
 :3287
 :6.1,III
 :GHS07
 :10361-37-2
 :BaCl₂
 :MFCD00003445