Sodium thiosulphate 0.05 mol/l (0.05 N) in aqueous solution
Supplier: Bernd Kraft
Jump to:
About this item
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
- CAS Number:7772-98-7
- Formula:Na₂S₂O₃
- MDL Number:MFCD00003499
- Molecular Weight:158.11 g/mol
- Storage Temperature:Ambient
- EINECS:231-867-5
5 Options Available Below
Product Details & Documents
Synonyms: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
- CAS Number:7772-98-7
- Formula:Na₂S₂O₃
- MDL Number:MFCD00003499
- Molecular Weight:158.11 g/mol
- Storage Temperature:Ambient
- EINECS:231-867-5
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
This product has been blocked by your organization. Please contact your purchasing department for more information



