Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 4 M NaOH (4N) volumetric solution, Fluka™
Corrosive
Sodium hydroxide 4 M NaOH (4N) volumetric solution, Fluka™
Catalog # 35274-1L
CAS Number:  
Corrosive
Sodium hydroxide 4 M NaOH (4N) volumetric solution, Fluka™
Catalog # 35274-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:105…140 °C
Melting Pt:–10–14 °C
Density:1.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548