Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 2 M NaOH (2N) ≥99.5% volumetric solution, Fluka™
Corrosive
Sodium hydroxide 2 M NaOH (2N) ≥99.5% volumetric solution, Fluka™
Certificates
Corrosive
Sodium hydroxide 2 M NaOH (2N) ≥99.5% volumetric solution, Fluka™
Certificates

Specification Test Results

  • Assay:
    1.996 - 2.004 mol/l
  • Calibrated Against:
    potassium hydrogen phthalate
  • Vapor Pressure:
    3 mmHg ( 37 °C)

Specification Test Results

Related Information

Frequently Bought Together