Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution
Catalog # HACH67132
Supplier:  Hach
CAS Number:  
Sodium hydroxide 0.01 mol/l (0.01 N) in aqueous solution
Catalog # HACH67132
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548