Order Entry
Belgium
ContactUsLinkComponent
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution volumetric solution
Corrosive
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution volumetric solution
Catalog # REGEKOH20235
Supplier:  REAGECON
CAS Number:  
Corrosive
Potassium hydroxide 0.23 mol/l (0.23 N) in aqueous solution volumetric solution
Catalog # REGEKOH20235
Supplier:  REAGECON
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:1814
ADR:8,II
 :GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553