VWR® ARISTAR®, Zirconium Standard Solution, 1000 mg/L Zr in dil. Nitric Acid (from ZrO(NO₃)₂), Standard for ICP
Catalog # 456402S
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:Nitric acid dil.
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Zirconium
- Symbol:Zr
Specifications
About this item
ZrO(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.