Order Entry
Belgium
ContactUsLinkComponent
 
VWR® ARISTAR®, Strontium Standard Solution, 10000 mg/L Sr in dil. Nitric Acid (from Sr(NO₃)₂), Standard for ICP
  456072U
 :  
undefined
VWR® ARISTAR®, Strontium Standard Solution, 10000 mg/L Sr in dil. Nitric Acid (from Sr(NO₃)₂), Standard for ICP
  456072U
 :  

 

  • Concentration:
    10000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Strontium
  • Symbol:
    Sr

 

 

Sr(NO₃)₂ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.