Order Entry
Belgium
ContactUsLinkComponent
Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Catalog # 32038-1EA
CAS Number:  
Sodium chloride concentrate for dilution to 1L, 0.1 M NaCl (0.1N) volumetric solution, Fluka™
Catalog # 32038-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :58.44 g/mol
Density:1.00 g/cm³
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477