Order Entry
Belgium
ContactUsLinkComponent
 
VWR®, Scandium standard solution, 1000 mg/L Sc in dil. nitric acid (from Sc2O3) ARISTAR® standard for ICP
  455962S
 :  
undefined
VWR®, Scandium standard solution, 1000 mg/L Sc in dil. nitric acid (from Sc2O3) ARISTAR® standard for ICP
  455962S
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Scandium
  • Symbol:
    Sc

 

 

Sc₂O₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.