Order Entry
Belgium
ContactUsLinkComponent
 
VWR®, Mercury standard solution, 1,000 mg/l Hg in dil. nitric acid (from Hg) ARISTAR® standard for ICP
  455662J
 :  
undefined
VWR®, Mercury standard solution, 1,000 mg/l Hg in dil. nitric acid (from Hg) ARISTAR® standard for ICP
  455662J
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Mercury
  • Symbol:
    Hg

 

 

Hg in HNO₃ 10%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.