VWR® ARISTAR®, Platinum Standard Solution, 1000 mg/L Pt in 10% Hydrochloric Acid (from Pt), Standard for ICP
N° de catalogue 455802D
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Platinum
- Symbol:Pt
Specifications
About this item
Pt in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.