VWR® ARISTAR®, Phosphorus Standard Solution, 1000 mg/L P in Water (from (NH₄)PO₄H₂), Standard for ICP
Catalog # 455782Q
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Plastic bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Phosphorus
- Symbol:P
Specifications
About this item
NH₄H₂PO₄ in H₂O.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.