VWR® ARISTAR®, Palladium Standard Solution, 1000 mg/l Pd in 10% Hydrochloric Acid (from Pd), Standard for ICP
Catalog # 455762M
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:10% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Palladium
- Symbol:Pd
Specifications
About this item
Pd in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.