VWR®, Neodymium standard solution, 1000 mg/L Nd in dil. nitric acid (from Nd₂O₃) ARISTAR® standard for ICP
Catalog # 455702A
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Neodymium
- Symbol:Nd
Specifications
About this item
Nd₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.