VWR®, Magnesium standard solution, 1000 mg/L Mg in dil. nitric acid AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
Certificates
About this item
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together