Order Entry
Belgium
ContactUsLinkComponent
 
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid (from Lu₂O₃) Certipur® standard for ICP, Supelco®
  1.70330.0100
 :  Merck
 :  
undefined
Lutetium standard solution, 1000 mg/L Lu in dil. nitric acid (from Lu₂O₃) Certipur® standard for ICP, Supelco®
  1.70330.0100
 :  Merck
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Lutetium
  • Symbol:
    Lu

 

 

 : Traceable to SRM from NIST.