Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li in dil. Nitric Acid (from Li₂CO₃), Standard for ICP
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li in dil. Nitric Acid (from Li₂CO₃), Standard for ICP
Catalog # 455582K
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li in dil. Nitric Acid (from Li₂CO₃), Standard for ICP
Catalog # 455582K
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item

Li₂CO₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.