Order Entry
Belgium
ContactUsLinkComponent
Copper standard solution, 10000 mg/L Cu in dil. nitric acid (from Cu(NO₃₃)₂) Certipur® standard for ICP, Supelco®
Copper standard solution, 10000 mg/L Cu in dil. nitric acid (from Cu(NO₃₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70378.0100
Supplier:  Merck
CAS Number:  
Copper standard solution, 10000 mg/L Cu in dil. nitric acid (from Cu(NO₃₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70378.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item

Certifications: Traceable to SRM from NIST.