ARISTAR®VWR® , Potassium Standard Solution, 10000 mg/l K in dil. Nitric Acid (from KNO3), Standard for ICP
Catalog # 455832J
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:10000mg/L
- Matrix:2-5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Potassium
- Symbol:K
Specifications
About this item
KNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.