Order Entry
Belgium
ContactUsLinkComponent
VWR® ARISTAR®, Potassium Standard Solution, 1000 mg/L K in dilute Nitric Acid (from KNO₃), Standard for ICP
VWR® ARISTAR®, Potassium Standard Solution, 1000 mg/L K in dilute Nitric Acid (from KNO₃), Standard for ICP
Catalog # 455822H
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® ARISTAR®, Potassium Standard Solution, 1000 mg/L K in dilute Nitric Acid (from KNO₃), Standard for ICP
Catalog # 455822H
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Potassium
  • Symbol:
    K

Specifications

About this item

KNO₃ in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.