Order Entry
Belgium
ContactUsLinkComponent
VWR®, Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) ARISTAR® standard for ICP
VWR®, Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) ARISTAR® standard for ICP
Catalog # 455422S
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) ARISTAR® standard for ICP
Catalog # 455422S
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Gold
  • Symbol:
    Au

Specifications

About this item

Au in HCl 10%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.