VWR®, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid (from Eu2O3), Standard for ICP
Catalog # 455342T
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Europium
- Symbol:Eu
Specifications
About this item
Eu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.