Order Entry
Belgium
ContactUsLinkComponent
 
VWR® ARISTAR®, Iron Standard Solution, 1000 mg/L Fe in dil. Nitric Acid (from Fe), Standard for ICP
  455522V
 :  
undefined
VWR® ARISTAR®, Iron Standard Solution, 1000 mg/L Fe in dil. Nitric Acid (from Fe), Standard for ICP
  455522V
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    Nitric acid dil.
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Iron
  • Symbol:
    Fe

 

 

Fe in HNO₃ 2 to 5%.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.