Order Entry
Belgium
ContactUsLinkComponent
Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from Ca(NO₃)₂) Certipur® standard for ICP, Supelco®
Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from Ca(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70373.0100
Supplier:  Merck
CAS Number:  
Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from Ca(NO₃)₂) Certipur® standard for ICP, Supelco®
Catalog # 1.70373.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2-3% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

Certifications: Traceable to SRM from NIST.