Order Entry
Belgium
ContactUsLinkComponent
 
Calcium standard solution, 1.000 mg/l Ca in dil. nitric acid (from Ca) ARISTAR® standard for ion chromatography
Catalog # 458202U
Supplier:  VWR Chemicals
CAS Number:  
undefined
Calcium standard solution, 1.000 mg/l Ca in dil. nitric acid (from Ca) ARISTAR® standard for ion chromatography
Catalog # 458202U
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O/tr HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item

Ca in dilute HNO₃.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.