VWR® ARISTAR®, Arsenic Standard Solutions for ICP
Catalog # 455042K
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Arsenic
- Symbol:As
Specifications
About this item
As in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.