Order Entry
Belgium
ContactUsLinkComponent
 
VWR® ARISTAR®, Sulphur Standard Solution, 1000 mg/l S in Water (from (NH₄)₂SO₄), Standard for ICP
  456082W
 :  
undefined
VWR® ARISTAR®, Sulphur Standard Solution, 1000 mg/l S in Water (from (NH₄)₂SO₄), Standard for ICP
  456082W
 :  

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Sulphur
  • Symbol:
    S

 

 

(NH₄)₂SO₄ in H₂O.

Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.

Supplied with certificate of analysis.