Nitrite standard solution, 1000 mg/L NO₂- in Water (from NaNO₂) Certipur®, Supelco®
Catalog # 1.19899.0500
Supplier: Merck
CAS Number:
Specifications
- Concentration:1000 mg/L
- Pack type:Plastic bottle
- Pk:500 mL
- Description:Nitrite standard solution
- M-Clarity quality level:MQ100
- Contents:NaNO₂ in H₂O